Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-1-Boc-3-aminopyrrolidine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate(3R)-(+)-1-Boc-3-aminopyrrolidine(3R)-(+)-3-Amino-1-Boc-pyrrolidine(3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine(3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine

H28188-03 H28188-06
AAH28188-03EA 143.99 USD InStock
AAH28188-03 AAH28188-06
(R)-(+)-1-Boc-3-aminopyrrolidine 97%
(R)-(+)-1-Boc-3-aminopyrrolidine


Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Boiling Pt: 243…244 °C
Density: 1.098
Flash Pt: 91 °C(196 °F)
Storage Temperature: Ambient
MDL Number: MFCD03419272
CAS Number: 147081-49-0
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty