Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(4-Methoxyphenyl)propylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester

H28557-06 H28557-MD H28557-03
AAH28557-03EA 337.99 USD InStock
AAH28557-03 AAH28557-MD AAH28557-06
3-(4-Methoxyphenyl)propylboronic acid pinacol ester 97%
3-(4-Methoxyphenyl)propylboronic acid pinacol ester


Formula: C₁₆H₂₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD09953503
CAS Number: 1073371-72-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty