Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Benzylbenzeneboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals

H28682-06 H28682-03
AAH28682-06EA 359.99 USD InStock
AAH28682-06 AAH28682-03
3-Benzylbenzeneboronic acid pinacol ester 97%
3-Benzylbenzeneboronic acid pinacol ester


Formula: C₁₉H₂₃BO₂
MW: 294.20 g/mol
Melting Pt: 68…69 °C
Storage Temperature: Ambient
MDL Number: MFCD09266179
CAS Number: 1073355-05-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty