Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-5-fluoropyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

H30170-03 H30170-06
AAH30170-03EA 141.99 USD InStock
AAH30170-03 AAH30170-06
3-Chloro-5-fluoropyridine 98%
3-Chloro-5-fluoropyridine


Formula: C₅H₃ClFN
Melting Pt: 25…27 °C
Flash Pt: 41 °C(106 °F)
Storage Temperature: Ambient
MDL Number: MFCD05663707
CAS Number: 514797-99-0
UN: 1325
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty