Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxy-trans-β-styrylboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (E)-2-(4-Methoxystyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H30373-MD H30373-06 H30373-03
AAH30373-03EA 171.46 USD InStock
AAH30373-03 AAH30373-MD AAH30373-06
4-Methoxy-trans-β-styrylboronic acid pinacol ester 98%
4-Methoxy-trans-beta-styrylboronic acid pinacol ester


Formula: C₁₅H₂₁BO₃
MW: 260.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038966
CAS Number: 149777-83-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty