Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Hydroxybutyric acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-3-Hydroxybutanoic acid3-Hydroxybutyric acidDL-3-Hydroxybutyric Acid

H30426-14 H30426-06
AAH30426-06EA 216.99 USD InStock
AAH30426-06 AAH30426-14
(±)-3-Hydroxybutyric acid 98%
(±)-3-Hydroxybutyric acid


Formula: C₄H₈O₃
MW: 104.11 g/mol
Melting Pt: 36…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00004546
CAS Number: 300-85-6
EINECS: 206-099-9
Merck Index: 12,04861
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty