Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-3-Amino-1-boc-piperidine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S)-(+)-3-Amino-1-BOC-piperidine(S)-tert-Butyl 3-aminopiperidine-1-carboxylate

H30538-03 H30538-06 H30538-MD
AAH30538-MDEA 85.13 USD InStock
AAH30538-MD AAH30538-06 AAH30538-03
(S)-(+)-3-Amino-1-boc-piperidine 97%
(S)-(+)-3-Amino-1-Boc-piperidine


Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094718
CAS Number: 625471-18-3
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty