Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-6-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 6-Fluoro-2-pyridylamine 6-Fluoropyridin-2-amine

H30629-06 H30629-03
AAH30629-06EA 324.99 USD InStock
AAH30629-06 AAH30629-03
2-Amino-6-fluoropyridine 97%
2-Amino-6-fluoropyridine


Formula: C₅H₅FN₂
MW: 112.11 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD04114200
CAS Number: 1597-32-6
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty