Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Boc-amino)pyridine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-Butyl pyridin-2-ylcarbamatetert-Butyl N-(2-pyridyl)carbamate

H30741-06 H30741-03
AAH30741-03EA 93.69 USD InStock
AAH30741-03 AAH30741-06
2-(Boc-amino)pyridine 95%
2-(Boc-amino)pyridine


Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD03411622
CAS Number: 38427-94-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty