Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Boc-amino)pyridine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Boc-Amino)pyridine

H30820-06 H30820-14
AAH30820-06EA 83.89 USD InStock
AAH30820-06 AAH30820-14
4-(Boc-amino)pyridine 95%
4-(Boc-amino)pyridine


Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD02179232
CAS Number: 98400-69-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty