Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

H30924-06 H30924-03
AAH30924-03EA 109.99 USD InStock
AAH30924-03 AAH30924-06
2-Bromo-6-fluoropyridine 97%
2-Bromo-6-fluoropyridine


Formula: C₅H₃BrFN
Melting Pt: 30…34 °C
Storage Temperature: Ambient
MDL Number: MFCD03095258
CAS Number: 144100-07-2
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty