Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodoisonicotinic acid 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Iodopyridine-4-carboxylic acid

H31002-03 H31002-MD
AAH31002-MDEA 58.05 USD InStock
AAH31002-MD AAH31002-03
2-Iodoisonicotinic acid 95%
2-Iodoisonicotinic acid


Formula: C₆H₄INO₂
Melting Pt: ca 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06800803
CAS Number: 58481-10-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty