Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Formylglycine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: For-Gly-OH

H31053-03 H31053-06
AAH31053-06EA 111.99 USD InStock
AAH31053-06 AAH31053-03
N-Formylglycine 98%
N-Formylglycine


Formula: C₃H₅NO₃
MW: 103.08 g/mol
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00037360
CAS Number: 2491-15-8
EINECS: 219-649-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty