Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Benzyloxy-2-fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Benzyloxy)-2-fluorobenzoic acid

H31488-06 H31488-03
AAH31488-06EA 192.99 USD InStock
AAH31488-06 AAH31488-03
4-Benzyloxy-2-fluorobenzoic acid 98%
4-Benzyloxy-2-fluorobenzoic acid


Formula: C₁₄H₁₁FO₃
MW: 246.24 g/mol
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD12407082
CAS Number: 114045-96-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty