Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethylthio)benzylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

H31515-03 H31515-06
AAH31515-03EA 132.99 USD InStock
AAH31515-03 AAH31515-06
4-(Trifluoromethylthio)benzylamine 95%
4-(Trifluoromethylthio)benzylamine


Formula: C₈H₈F₃NS
Boiling Pt: 224…225 °C
Density: 1.310
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190129
CAS Number: 128273-56-3
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty