Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-methoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H31604-03 H31604-06
AAH31604-06EA 280.99 USD InStock
AAH31604-06 AAH31604-03
2-Bromo-6-methoxybenzoic acid 98%
2-Bromo-6-methoxybenzoic acid


Formula: C₈H₇BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00800732
CAS Number: 31786-45-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty