Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-2-hydroxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H31705-03 H31705-MD H31705-06
AAH31705-03EA 333.99 USD InStock
AAH31705-03 AAH31705-MD AAH31705-06
3-Fluoro-2-hydroxypyridine 97%
3-Fluoro-2-hydroxypyridine


Formula: C₅H₄FNO
MW: 113.09 g/mol
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD03095042
CAS Number: 1547-29-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty