Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrrolidin-2-ylacetic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Pyrrolidin-2-yl-acetic acid hydrochloride

H31749-MD H31749-03
AAH31749-MDEA 79.89 USD InStock
AAH31749-MD AAH31749-03
Pyrrolidin-2-ylacetic acid hydrochloride 97%
Pyrrolidin-2-ylacetic acid hydrochloride


Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD11506301
CAS Number: 71985-79-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty