Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Bromopicolinamide 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Bromopyridine-2-carboxamide

H31811-03 H31811-06
AAH31811-03EA 61.59 USD InStock
AAH31811-03 AAH31811-06
6-Bromopicolinamide 97%
6-Bromopicolinamide


Formula: C₆H₅BrN₂O
MW: 201.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234154
CAS Number: 25194-52-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty