Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethoxy-2,4-difluorobenzoic acid 97%, JRD

Supplier: Thermo Scientific Chemicals
Warning

H31812-03 H31812-06
AAH31812-03EA 94.19 USD InStock
AAH31812-03 AAH31812-06
3-Ethoxy-2,4-difluorobenzoic acid 97%, JRD
3-Ethoxy-2,4-difluorobenzoic acid


Formula: C₉H₈F₂O₃
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD09258723
CAS Number: 1017779-88-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty