Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-methylbenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Bromo-2-methylbenzene-1-sulfonyl chloride4-Bromo-2-methylbenzene-1-sulphonyl chloride

H31835-09 H31835-03
AAH31835-03EA 49.69 USD InStock
AAH31835-03 AAH31835-09
4-Bromo-2-methylbenzenesulfonyl chloride 97%
4-Bromo-2-methylbenzenesulphonyl chloride


Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Boiling Pt: 164 °C/15 mm
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00219927
CAS Number: 139937-37-4
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty