Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-5-methoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H31960-03 H31960-06
AAH31960-06EA 162.99 USD InStock
AAH31960-06 AAH31960-03
3-Chloro-5-methoxybenzoic acid 98%
3-Chloro-5-methoxybenzoic acid


Formula: C₈H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD01317994
CAS Number: 82477-67-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty