Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluoro-6-nitrophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

H32153-03 H32153-06
AAH32153-06EA 190.99 USD InStock
AAH32153-06 AAH32153-03
4-Bromo-2-fluoro-6-nitrophenol 98%
4-Bromo-2-fluoro-6-nitrophenol


Formula: C₆H₃BrFNO₃
MW: 236.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013372
CAS Number: 320-76-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty