Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Bromoacetyl)pyridine hydrobromide

H32285-14 H32285-06 H32285-03
AAH32285-06EA 277.99 USD InStock
AAH32285-06 AAH32285-14 AAH32285-03
2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide 98%
2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide


Formula: C₇H₇Br₂NO
MW: 280.95
Melting Pt: ca 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02681893
CAS Number: 5349-17-7
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty