Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N-methylacetamide 97%

Supplier: Thermo Scientific Chemicals
Danger

H32362-06 H32362-14
AAH32362-14EA 272.99 USD InStock
AAH32362-14 AAH32362-06
2-Chloro-N-methylacetamide 97%
2-Chloro-N-methylacetamide


Formula: C₃H₆ClNO
MW: 107.54 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00018913
CAS Number: 96-30-0
EINECS: 202-497-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty