Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-α,α,α-trifluoro-m-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Amino-3-(trifluoromethyl)benzoic acid

H32487-09 H32487-03
AAH32487-03EA 43.19 USD InStock
AAH32487-03 AAH32487-09
4-Amino-α,α,α-trifluoro-m-toluic acid 98%
4-Amino-α,α,α-trifluoro-m-toluic acid


Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 212…215 °C
Storage Temperature: Ambient
MDL Number: MFCD03407960
CAS Number: 400-76-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty