Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-bromo-4,4,4-trifluorobutyrate 97%

Supplier: Thermo Scientific Chemicals
Danger

H32572-03 H32572-MD
AAH32572-MDEA 70.79 USD InStock
AAH32572-MD AAH32572-03
Ethyl-2-bromo-4,4,4-trifluorobutyrate 97%
Ethyl-2-bromo-4,4,4-trifluorobutyrate


Formula: C₆H₈BrF₃O₂
MW: 249.03 g/mol
Boiling Pt: 67 °C/18 mm
Storage Temperature: Ambient
MDL Number: MFCD10687235
CAS Number: 367-33-9
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty