Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-ethylamine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-Butyl N-ethylcarbamatetert-Butyl ethylcarbamate

H32575-03 H32575-06
AAH32575-06EA 239.99 USD InStock
AAH32575-06 AAH32575-03
N-Boc-ethylamine 97%
N-Boc-ethylamine


Formula: C₇H₁₅NO₂
MW: 145.20 g/mol
Melting Pt: 38…41 °C
Storage Temperature: Ambient
MDL Number: MFCD12923020
CAS Number: 38267-76-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty