Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene 97%

Supplier: Thermo Scientific Chemicals
Warning

H32580-03 H32580-09
AAH32580-09EA 227.99 USD InStock
AAH32580-09 AAH32580-03
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene 97%
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene


Formula: C₇H₂BrCl₂F₃O
MW: 309.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412210
CAS Number: 118754-55-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty