Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-2-nitrobenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

H32610-03 H32610-09
AAH32610-09EA 527.99 USD InStock
AAH32610-09 AAH32610-03
4-Fluoro-2-nitrobenzonitrile 97%
4-Fluoro-2-nitrobenzonitrile


Formula: C₇H₃FN₂O₂
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00277447
CAS Number: 80517-21-1
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty