Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylpyridin-3-ol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Hydroxy-5-picoline3-Hydroxy-5-methylpyridine

H32658-06 H32658-MD H32658-03
AAH32658-06EA 1003.99 USD InStock
AAH32658-06 AAH32658-MD AAH32658-03
5-Methylpyridin-3-ol 97%
5-Methylpyridin-3-ol


Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00661297
CAS Number: 42732-49-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty