Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxythiobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Hydroxybenzothioamide

H32780-14 H32780-06
AAH32780-14EA 374.99 USD InStock
AAH32780-14 AAH32780-06
4-Hydroxythiobenzamide 98%
4-Hydroxythiobenzamide


Formula: C₇H₇NOS
MW: 153.2 g/mol
Melting Pt: 181…185 °C
Storage Temperature: Ambient
MDL Number: MFCD04973332
CAS Number: 25984-63-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty