Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-2-iodo-3-hydroxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Chloro-2-iodopyridin-3-ol

H33127-03 H33127-09
AAH33127-03EA 76.29 USD InStock
AAH33127-03 AAH33127-09
6-Chloro-2-iodo-3-hydroxypyridine 98%
6-Chloro-2-iodo-3-hydroxypyridine


Formula: C₅H₃ClIO
MW: 255.44 g/mol
Melting Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD07636749
CAS Number: 188057-26-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty