Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-1,3-benzothiazol-6-amine 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Amino-2-methyl-1,3-benzothiazole

H33249-06 H33249-03
AAH33249-03EA 59.19 USD InStock
AAH33249-03 AAH33249-06
2-Methyl-1,3-benzothiazol-6-amine 96%
2-Methyl-1,3-benzothiazol-6-amine


Formula: C₈H₈N₂S
MW: 164.23 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00160076
CAS Number: 2941-62-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty