Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Trifluoromethyl)cyclohexanone 97%

Supplier: Thermo Scientific Chemicals
Warning

H33423-MD H33423-03 H33423-MD H33423-03
AAH33423-MDEA 49.99 USD InStock
CAAAH33423-MD AAH33423-MD AAH33423-03 CAAAH33423-03
2-(Trifluoromethyl)cyclohexanone 97%
2-(Trifluoromethyl)cyclohexanone


Formula: C₇H₉F₃O
MW: 166.14
Boiling Pt: 71…73/12 mm
Storage Temperature: Ambient
MDL Number: MFCD00102125
UN: 1224
ADR: "3",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty