Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine 96%

Supplier: Thermo Scientific Chemicals
Warning

H33439-03 H33439-06 H33439-03 H33439-06
AAH33439-06EA 311.99 USD InStock
AAH33439-06 CAAAH33439-06 CAAAH33439-03 AAH33439-03
4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine 96%
4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine


Formula: C₁₁H₁₁ClN₂S
MW: 238.733 g/mol
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD02333821
CAS Number: 137438-23-4
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty