Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Formyl-L-leucine 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Formyl-Leu-OH

H33564-06 H33564-06 H33564-03 H33564-03
AAH33564-03EA 45.59 USD InStock
CAAAH33564-03 AAH33564-03 AAH33564-06 CAAAH33564-06
N-Formyl-L-leucine 90%, Technical Grade
N-Formyl-L-leucine


Formula: C₇H₁₃NO₃
MW: 159.19 g/mol
Melting Pt: 142…145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00055861
CAS Number: 6113-61-7
EINECS: 228-080-4
Order Now
Specification Test Results
Assay Minimum 90

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty