Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Cyclohexylbenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Cyclohexylbenzene-1-sulfonyl chloride4-Cyclohexylbenzene-1-sulphonyl chloride

H33616-09 H33616-03 H33616-09 H33616-03
AAH33616-09EA 547.99 USD InStock
AAH33616-09 CAAAH33616-09 CAAAH33616-03 AAH33616-03
4-Cyclohexylbenzenesulfonyl chloride 97%
4-Cyclohexylbenzenesulfonyl chloride


Formula: C₁₂H₁₅ClO₂S
MW: 258.76 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00625713
CAS Number: 56354-57-5
UN: 3261
ADR: "8",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty