Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl chlorophosphate 97+%

Supplier: Thermo Scientific Chemicals
Danger

H33820-22 H33820-22 H33820-14 H33820-14
AAH33820-14EA 65.77 USD InStock
AAH33820-14 CAAAH33820-14 AAH33820-22 CAAAH33820-22
Diethyl chlorophosphate 97+%
Diethyl chlorophosphate


Formula: C₄H₁₀ClO₃P
Boiling Pt: 60 °C/2 mm
Density: 1.194
Flash Pt: 61 °C(142 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009075
CAS Number: 814-49-3
EINECS: 212-369-4
UN: 2927
ADR: "6.1",II
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty