Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Chloro-2-hydrazinyl-5-(trifluoromethyl)pyridine

H33850-06 H33850-03 H33850-06 H33850-03
AAH33850-03EA 79.79 USD InStock
CAAAH33850-03 AAH33850-03 AAH33850-06 CAAAH33850-06
3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine 97%
3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine


Formula: C₆H₅ClF₃N₃
MW: 211.58 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00067865
CAS Number: 89570-82-1
UN: 2811
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty