Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-nitro-3,4-dihydro-2H-1,4-benzoxazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H33900-03 H33900-MD
AAH33900-03EA 93.39 USD InStock
AAH33900-03 AAH33900-MD
6-nitro-3,4-dihydro-2H-1,4-benzoxazine 97%
6-nitro-3,4-dihydro-2H-1,4-benzoxazine


Formula: C₈H₈N₂O₃
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD07698591
CAS Number: 28226-22-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty