Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pentafluorophenyl 11-(triethoxysilyl)undecanoate

H34037-03 H34037-MD H34037-MD H34037-03
AAH34037-03EA 329.99 USD InStock
AAH34037-03 CAAAH34037-MD CAAAH34037-03 AAH34037-MD
10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane 95%
10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane


Formula: C₂₃H₃₅F₅O₅Si
MW: 514.61 g/mol
Boiling Pt: 155…160 °C/0.04 mm
Storage Temperature: Ambient
MDL Number: MFCD13181916
CAS Number: 1197981-08-0
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty