Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4-(2-thienyl)benzoic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

H34049-03 H34049-MD H34049-03 H34049-MD
AAH34049-03EA 193.51 USD InStock
CAAAH34049-MD AAH34049-03 CAAAH34049-03 AAH34049-MD
2-Amino-4-(2-thienyl)benzoic acid 96%
2-Amino-4-(2-thienyl)benzoic acid


Formula: C₁₁H₉NO₂S
MW: 219.258 g/mol
Melting Pt: 229…232 °C
Storage Temperature: Ambient
MDL Number: MFCD07357339
CAS Number: 948006-04-0
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty