Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Difluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H34188-MD H34188-MD H34188-03 H34188-03
AAH34188-03EA 317.99 USD InStock
CAAAH34188-03 CAAAH34188-MD AAH34188-03 AAH34188-MD
3-(Difluoromethyl)aniline 97%
3-(Difluoromethyl)aniline


Formula: C₇H₇F₂N
MW: 143.137 g/mol
Boiling Pt: 82 °C/4 mm
Storage Temperature: Ambient
MDL Number: MFCD18384830
CAS Number: 368-99-0
UN: 2810
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty