Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-iodonicotinonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H34191-MD H34191-03 H34191-06
AAH34191-MDEA 81.52 USD InStock
AAH34191-MD AAH34191-03 AAH34191-06
2-Amino-5-iodonicotinonitrile 97%
2-Amino-5-iodonicotinonitrile


Formula: C₆H₄IN₃
Melting Pt: 193…197 °C
Storage Temperature: Ambient
MDL Number: MFCD20040120
CAS Number: 1347815-41-1
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty