Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Thienyl)benzenesulfonyl chloride 96%

Supplier: Thermo Scientific Chemicals
Danger

H34399-MD H34399-06 H34399-03
AAH34399-MDEA 42.79 USD InStock
AAH34399-MD AAH34399-06 AAH34399-03
4-(2-Thienyl)benzenesulfonyl chloride 96%
4-(2-Thienyl)benzenesulfonyl chloride


Formula: C₁₀H₇ClO₂S₂
MW: 258.75 g/mol
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD11655569
CAS Number: 209592-91-6
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty