Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methyl-3-indolemethylamine 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Aminomethyl)-1-methylindole

H34418-MD H34418-03 H34418-06
AAH34418-03EA 187.99 USD InStock
AAH34418-03 AAH34418-MD AAH34418-06
1-Methyl-3-indolemethylamine 96%
1-Methyl-3-indolemethylamine


Formula: C₁₀H₁₂N₂
MW: 160.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657101
CAS Number: 19293-60-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty