Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Eicosanethiol 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Eicosyl MercaptanIcosyl Mercaptan

H34498-MD H34498-03
AAH34498-MDEA 86.39 USD InStock
AAH34498-MD AAH34498-03
1-Eicosanethiol 98%
1-Eicosanethiol


Formula: C₂₀H₄₂S
MW: 314.62 g/mol
Melting Pt: 38…41 °C
Storage Temperature: Ambient
MDL Number: MFCD07779406
CAS Number: 13373-97-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty