Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Imidazo[1,2-a]pyrid-2-yl)aniline 95%

Supplier: Thermo Scientific Chemicals
Warning

H35666-03 H35666-06 H35666-03 H35666-06
AAH35666-03EA 160.99 USD InStock
AAH35666-03 AAH35666-06 CAAAH35666-06 CAAAH35666-03
4-(Imidazo[1,2-a]pyrid-2-yl)aniline 95%
4-(Imidazo[1,2-a]pyrid-2-yl)aniline


Formula: C₁₃H₁₁N₃
MW: 209.252 g/mol
Melting Pt: ca. 230°C dec. (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00107394
CAS Number: 139705-74-1
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty