Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one 95%

Supplier: Thermo Scientific Chemicals
Danger

H36528-03 H36528-06
AAH36528-06EA 596.99 USD InStock
AAH36528-06 AAH36528-03
1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one 95%
1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one


Formula: C₅H₉BrN₄O
Boiling Pt: 223 °C
Density: 1.777
Flash Pt: 89 °C(192 °F)
Storage Temperature: Ambient
MDL Number: MFCD16251259
CAS Number: 84501-67-7
EINECS: 282-986-4
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty